Search
Search

KIF25-AS1| Gene Symbol: KIF25-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103750.1 | 2 | 509 | |||
| GenBank mRNA | AB016899.1 | - | 1 | 813 | ||
| AB016900.1 | - | 2 | 509 | |||
| BC112215.1 | - | 1 | 244 | |||
| BC143766.1 | - | 1 | 244 | 9052280 | ||