Search
Search

SNX12| Gene Symbol: SNX12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256185.1 | NP_001243114.1 | 2, 3 | 322 | ||
| NM_001256187.1 | NP_001243116.1 | 1, 2 | 298 | |||
| NM_013346.3 | NP_037478.2 | 1, 2 | 298 | |||
| GenBank mRNA | AF171229.1 | - | 1, 2 | 175 | ||
| AK312977.1 | - | 1, 2 | 246 | |||
| BC020559.1 | - | 1, 2 | 190 | |||
| BC080535.1 | - | 1, 2 | 173 | |||
| BC103847.2 | - | 1, 2 | 188 | 40002749 | ||
| BC103848.1 | - | 1, 2 | 188 | 40002752 | ||
| BM805683.1 | - | 345 | ||||
| BT007203.1 | - | 1, 2 | 156 | |||
| DA736880.1 | - | 2, 3 | 322 | |||
| DB251077.1 | - | 1, 2 | 254 | |||
| DB482683.1 | - | 1, 2 | 298 | |||