Search
Search

NOSIP| Gene Symbol: NOSIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270960.1 | NP_001257889.1 | 8 | 850 | ||
| NM_015953.4 | NP_057037.1 | 9 | 969 | |||
| XM_005258964.4 | XP_005259021.1 | 8 | 886 | |||
| XM_006723235.3 | XP_006723298.1 | 8 | 920 | |||
| XM_011527015.2 | XP_011525317.1 | 8 | 894 | |||
| XM_011527017.2 | XP_011525319.1 | 8 | 926 | |||
| XM_017026851.1 | XP_016882340.1 | 8 | 944 | |||
| XM_017026852.1 | XP_016882341.1 | 8 | 925 | |||
| GenBank mRNA | AF132959.1 | - | 8 | 835 | ||
| AK092444.1 | - | 6 | 2045 | |||
| BC009299.2 | - | 8 | 820 | 4125665 | ||
| BC010077.2 | - | 9 | 943 | 4309619 | ||
| BC011249.1 | - | 7 | 804 | 3452886 | ||