Search
Search

FAM198B| Gene Symbol: FAM198B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031700.2 | NP_001026870.2 | 2 | 1227 | ||
| NM_001128424.1 | NP_001121896.1 | 2 | 1227 | |||
| NM_016613.6 | NP_057697.2 | 2 | 1272 | |||
| GenBank mRNA | AF223468.1 | - | 1 | 214 | ||
| AK172805.1 | - | 2 | 1225 | |||
| AK304112.1 | - | 1 | 152 | |||
| AK315768.1 | - | 1 | 343 | |||
| AL832589.1 | - | 2 | 1098 | |||
| AL834177.1 | - | 2 | 723 | |||
| AY358785.1 | - | 2 | 1189 | |||
| BC017701.1 | - | 1 | 175 | |||
| BC043193.2 | - | 2 | 1227 | 5288748 | ||
| BC100016.1 | - | 1 | 111 | |||