Search
Search

FAM198B| Gene Symbol: FAM198B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031700.2 | NP_001026870.2 | 2 | 1060 | ||
| NM_001128424.1 | NP_001121896.1 | 2 | 1060 | |||
| NM_016613.6 | NP_057697.2 | 2 | 1105 | |||
| GenBank mRNA | AF223468.1 | - | 1 | 50 | ||
| AK172805.1 | - | 2 | 1058 | |||
| AK315768.1 | - | 1 | 176 | |||
| AL832589.1 | - | 2 | 931 | |||
| AL834177.1 | - | 2 | 557 | |||
| AY358785.1 | - | 2 | 1022 | |||
| BC043193.2 | - | 2 | 1060 | 5288748 | ||