Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MED15 SIRNA Assays ›
          Gene Symbol:
          MED15
          Gene Name:
          mediator complex subunit 15
          Gene Aliases:
          ARC105, CAG7A, CTG7A, PCQAP, TIG-1, TIG1, TNRC7
          Chromosome Location:
          Chr.22: 20507542 - 20587632 on Build GRCh38
          UniGene ID:
          Hs.517421
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS122391
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MED15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001003891.2 NP_001003891.1 16, 17 2261
          NM_001293234.1 NP_001280163.1 15, 16 2090
          NM_001293235.1 NP_001280164.1 15, 16 2130
          NM_001293236.1 NP_001280165.1 14, 15 1928
          NM_001293237.1 NP_001280166.1 15, 16 2333
          NM_015889.4 NP_056973.2 15, 16 2141
          XM_006724263.3 XP_006724326.1 15, 16 2006
          XM_006724264.3 XP_006724327.1 16, 17 2232
          XM_011530214.2 XP_011528516.1 17, 18 2305
          XM_011530215.2 XP_011528517.1 15, 16 2127
          XM_017028817.1 XP_016884306.1 15, 16 2203
          XM_017028818.1 XP_016884307.1 17, 18 2403
          XM_017028819.1 XP_016884308.1 16, 17 2169
          XM_017028820.1 XP_016884309.1 15, 16 2112
          XM_017028823.1 XP_016884312.1 15, 16 3384
          XM_017028824.1 XP_016884313.1 17, 18 2301
          GenBank mRNA AF056191.1 - 15, 16 2089
          AF328769.1 - 15, 16 1998
          AK074230.1 - 10, 11 1288
          AK074268.1 - 15, 16 1934
          AK090465.1 - 16, 17 2193
          AK124276.1 - 8, 9 2407
          AK124925.1 - 8, 9 1612
          AK126208.1 - 11, 12 2466
          AK294540.1 - 17, 18 2137
          AK294734.1 - 15, 16 2329
          AK297284.1 - 9, 10 1604
          AK297789.1 - 7, 8 865
          AK315540.1 - 15, 16 2053
          AL832151.1 - 8, 9 1276
          AL832923.1 - 5, 6 594
          BC003078.1 - 15, 16 2062
          BC007529.1 - 8, 9 914
          BC013985.2 - 15, 16 2044 3506327
          BC017110.1 - 12, 13 1889
          CR456537.1 - 14, 15 1820

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001108325, XM_006248702, XM_006248706, XM_008768891.

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          RNA polymerase II transcription cofactor activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0