Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRMT7 SIRNA Assays ›
Gene Symbol:
PRMT7
Gene Name:
protein arginine methyltransferase 7
Gene Aliases:
-
Chromosome Location:
Chr.16: 68310974 - 68358832 on Build GRCh38
UniGene ID:
Hs.640229
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS122956
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRMT7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001184824.1
NP_001171753.1
12
1427
NM_001290018.1
NP_001276947.1
14
2218
NM_019023.2
NP_061896.1
14
1712
XM_011523112.2
XP_011521414.1
13
1577
XM_011523113.2
XP_011521415.1
14
1704
XM_011523115.2
XP_011521417.1
13
1521
XM_011523116.2
XP_011521418.1
13
1577
XM_011523121.2
XP_011521423.1
11
1342
XM_011523124.2
XP_011521426.1
13
1630
XM_011523125.2
XP_011521427.1
12
1472
XM_011523126.2
XP_011521428.1
12
1476
XM_011523128.2
XP_011521430.1
13
1521
XM_011523131.2
XP_011521433.1
12
1451
XM_017023290.1
XP_016878779.1
14
1704
XM_017023291.1
XP_016878780.1
14
1644
XM_017023292.1
XP_016878781.1
13
1521
XM_017023293.1
XP_016878782.1
14
1704
XM_017023294.1
XP_016878783.1
14
1644
XM_017023295.1
XP_016878784.1
13
1577
XM_017023296.1
XP_016878785.1
14
1704
XM_017023297.1
XP_016878786.1
14
1704
XM_017023298.1
XP_016878787.1
12
1572
XM_017023299.1
XP_016878788.1
12
1572
XM_017023302.1
XP_016878791.1
12
1472
XM_017023303.1
XP_016878792.1
13
1630
XM_017023304.1
XP_016878793.1
14
1704
XM_017023305.1
XP_016878794.1
11
1342
XM_017023307.1
XP_016878796.1
13
1630
XM_017023308.1
XP_016878797.1
13
1644
XM_017023309.1
XP_016878798.1
12
1472
XM_017023310.1
XP_016878799.1
12
1496
XM_017023311.1
XP_016878800.1
12
1428
XM_017023313.1
XP_016878802.1
13
1619
XM_017023314.1
XP_016878803.1
12
1379
XM_017023315.1
XP_016878804.1
13
1619
XM_017023316.1
XP_016878805.1
12
1379
GenBank mRNA
AK001502.1
-
14
1644
AK022739.1
-
10
1139
AK056647.1
-
14
2200
AK097175.1
-
12
2154
AK304605.1
-
12
1350
BC000146.1
-
12
1453
2900965
Back To Top
More Information
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
spliceosomal snRNP assembly
regulation of gene expression by genetic imprinting
transcription, DNA-templated
regulation of transcription, DNA-templated
histone methylation
peptidyl-arginine methylation
peptidyl-arginine methylation, to symmetrical-dimethyl arginine
peptidyl-arginine methylation, to asymmetrical-dimethyl arginine
cell differentiation
histone arginine methylation
DNA methylation involved in gamete generation
regulation of protein binding
histone H4-R3 methylation
histone-arginine N-methyltransferase activity
S-adenosylmethionine-dependent methyltransferase activity
protein-arginine N-methyltransferase activity
[myelin basic protein]-arginine N-methyltransferase activity
protein-arginine omega-N monomethyltransferase activity
protein-arginine omega-N asymmetric methyltransferase activity
protein-arginine omega-N symmetric methyltransferase activity
histone binding
ribonucleoprotein complex binding
histone methyltransferase activity (H4-R3 specific)
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0