Search
Search

UGT1A1| Gene Symbol: UGT1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000463.2 | NP_000454.1 | 1 | 617 | ||
| GenBank mRNA | AK290834.1 | - | 1 | 621 | ||
| AY435136.1 | - | 1 | 602 | |||
| BC128414.1 | - | 1 | 618 | 40117598 | ||
| BC128415.1 | - | 1 | 617 | 40117599 | ||
| DQ364247.1 | - | 1 | 602 | |||
| JQ686667.1 | - | 1 | 602 | |||
| JQ699637.1 | - | 1 | 602 | |||
| JQ699638.1 | - | 1 | 602 | |||
| JQ699639.1 | - | 1 | 602 | |||
| JQ699640.1 | - | 1 | 602 | |||
| JQ699641.1 | - | 1 | 602 | |||
| JQ699642.1 | - | 1 | 602 | |||
| JQ699643.1 | - | 1 | 602 | |||
| JQ699644.1 | - | 1 | 602 | |||
| JQ699645.1 | - | 1 | 602 | |||
| JQ699646.1 | - | 1 | 602 | |||
| JQ699647.1 | - | 1 | 602 | |||
| JQ699649.1 | - | 1 | 602 | |||
| JQ699650.1 | - | 1 | 602 | |||
| M57899.1 | - | 1 | 617 | |||