Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UCKL1 SIRNA Assays ›
Gene Symbol:
UCKL1
Gene Name:
uridine-cytidine kinase 1 like 1
Gene Aliases:
UCK1L, URKL1
Chromosome Location:
Chr.20: 63939829 - 63959435 on Build GRCh38
UniGene ID:
Hs.504998
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS123622
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UCKL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001193379.1
NP_001180308.1
14
1729
NM_017859.3
NP_060329.2
14
1501
NR_126526.1
14
1478
XM_005260216.2
XP_005260273.1
14
1469
XM_006723806.2
XP_006723869.1
14
1475
XM_006723807.2
XP_006723870.1
14
1472
XM_006723808.1
XP_006723871.1
14
1527
XM_006723809.1
XP_006723872.1
14
1507
XM_006723810.1
XP_006723873.1
14
1504
XM_006723811.3
XP_006723874.1
14
3496
XM_011528868.2
XP_011527170.1
14
3499
XM_011528869.1
XP_011527171.1
15
4770
XM_011528870.1
XP_011527172.1
14
1489
XM_011528871.1
XP_011527173.1
14
1516
XM_011528872.1
XP_011527174.1
15
4759
XM_017027896.1
XP_016883385.1
14
1504
XM_017027898.1
XP_016883387.1
14
1501
XM_017027899.1
XP_016883388.1
14
1498
XM_017027900.1
XP_016883389.1
14
2350
XM_017027901.1
XP_016883390.1
14
2345
XM_017027902.1
XP_016883391.1
15
1707
XM_017027903.1
XP_016883392.1
14
1510
XM_017027904.1
XP_016883393.1
14
4431
XM_017027905.1
XP_016883394.1
14
4428
XM_017027906.1
XP_016883395.1
14
4425
XM_017027907.1
XP_016883396.1
10
1227
XM_017027908.1
XP_016883397.1
11
1130
GenBank mRNA
AJ605558.1
-
14
1447
AK000524.1
-
14
1470
AK222559.1
-
14
1472
AK223220.1
-
14
1470
AK303688.1
-
14
1681
BC033078.1
-
12
1488
4810505
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotide kinase
Gene Ontology Categories:
Function(s)
Process(es)
pyrimidine nucleobase metabolic process
viral process
phosphorylation
pyrimidine nucleoside salvage
UMP salvage
CTP salvage
uridine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0