Search
Search

FIGN| Gene Symbol: FIGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321825.1 | NP_001308754.1 | 2 | 2158 | ||
| NM_018086.3 | NP_060556.2 | 3 | 2328 | |||
| XM_011511389.2 | XP_011509691.1 | 4 | 2732 | |||
| XM_017004419.1 | XP_016859908.1 | 2 | 12265 | |||
| GenBank mRNA | AK001267.1 | - | 1 | 1815 | ||
| AK025747.1 | - | 1 | 620 | |||
| AK125324.1 | - | 3 | 2329 | |||
| BX649105.1 | - | 3 | 2326 | |||