Search
Search

CNOT11| Gene Symbol: CNOT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017546.4 | NP_060016.3 | 4, 5 | 1122 | ||
| GenBank mRNA | AF103798.1 | - | 4, 5 | 1113 | ||
| AK024221.1 | - | 3, 4 | 539 | |||
| BC018664.2 | - | 4, 5 | 1031 | |||
| BC064421.1 | - | 4, 5 | 1089 | 5752134 | ||