Search
Search

CNOT11| Gene Symbol: CNOT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017546.4 | NP_060016.3 | 3 | 861 | ||
| GenBank mRNA | AF103798.1 | - | 3 | 852 | ||
| AK024221.1 | - | 2 | 278 | |||
| BC018664.2 | - | 3 | 770 | |||
| BC064421.1 | - | 3 | 828 | 5752134 | ||