Search
Search

MAP7D1| Gene Symbol: MAP7D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286365.1 | NP_001273294.1 | 16 | 2924 | ||
| NM_001286366.1 | NP_001273295.1 | 18 | 2939 | |||
| NM_018067.4 | NP_060537.3 | 17 | 3038 | |||
| XM_005271027.1 | XP_005271084.1 | 16 | 2742 | |||
| XM_006710759.1 | XP_006710822.1 | 17 | 2850 | |||
| XM_006710761.1 | XP_006710824.1 | 17 | 2643 | |||
| GenBank mRNA | AB033013.2 | - | 16 | 2924 | ||
| AF218002.1 | - | 17 | 2262 | |||
| AK001212.1 | - | 9 | 1118 | |||
| AK095939.1 | - | 10 | 1282 | |||
| AK096341.1 | - | 18 | 2706 | |||
| AK309966.1 | - | 15 | 1930 | |||
| AL136547.1 | - | 9 | 949 | |||
| AY007099.1 | - | 9 | 1077 | |||
| BC003083.2 | - | 9 | 1185 | |||
| BC027334.1 | - | 7 | 710 | |||
| BC067256.1 | - | 14 | 2097 | |||
| BC082266.1 | - | 11 | 1460 | |||
| BC106053.1 | - | 17 | 2801 | 6514558 | ||
| BC128042.1 | - | 7 | 556 | |||