Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HHAT SIRNA Assays ›
Gene Symbol:
HHAT
Gene Name:
hedgehog acyltransferase
Gene Aliases:
MART2, SKI1, Skn
Chromosome Location:
Chr.1: 210327873 - 210676298 on Build GRCh38
UniGene ID:
Hs.58650
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS124910
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HHAT
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001122834.3
NP_001116306.1
5
461
NM_001170580.2
NP_001164051.1
5
516
NM_001170587.2
NP_001164058.1
4
393
NM_018194.5
NP_060664.2
5
527
XM_006711441.3
XP_006711504.1
6
588
XM_011509737.2
XP_011508039.1
6
588
XM_011509738.2
XP_011508040.1
4
346
XM_011509739.2
XP_011508041.1
5
513
XM_011509740.2
XP_011508042.1
6
717
XM_011509741.2
XP_011508043.1
5
461
XM_011509744.2
XP_011508046.1
4
521
XM_011509747.1
XP_011508049.1
3
311
XM_017001726.1
XP_016857215.1
4
562
XM_017001727.1
XP_016857216.1
4
562
XM_017001728.1
XP_016857217.1
5
671
XM_017001729.1
XP_016857218.1
5
392
XM_017001730.1
XP_016857219.1
4
474
XM_017001731.1
XP_016857220.1
4
280
XM_017001733.1
XP_016857222.1
3
434
XM_017001734.1
XP_016857223.1
4
468
XM_017001736.1
XP_016857225.1
4
368
GenBank mRNA
AK001586.1
-
5
481
AK295130.1
-
4
413
AK299200.1
-
3
311
AK302955.1
-
4
346
AK316190.1
-
4
347
BC039071.1
-
5
469
5177926
BC117130.1
-
4
316
40125681
DA336461.1
-
5
513
DB077466.1
-
5
451
DB080645.1
-
5
461
Back To Top
More Information
Panther Classification:
Molecular Function -
acyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
smoothened signaling pathway
multicellular organism development
protein palmitoylation
positive regulation of protein targeting to mitochondrion
GTP binding
O-acyltransferase activity
palmitoyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0