Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CSGALNACT1 SIRNA Assays ›
Gene Symbol:
CSGALNACT1
Gene Name:
chondroitin sulfate N-acetylgalactosaminyltransferase 1
Gene Aliases:
CSGalNAcT-1, ChGn, beta4GalNAcT
Chromosome Location:
Chr.8: 19404161 - 19757940 on Build GRCh38
UniGene ID:
Hs.613729
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS125048
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CSGALNACT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130518.1
NP_001123990.1
5
1736
NM_018371.4
NP_060841.5
5
1326
NR_024040.1
4
1118
XM_006716358.2
XP_006716421.1
4
1617
XM_006716359.2
XP_006716422.1
5
1986
XM_006716360.2
XP_006716423.1
6
2134
XM_006716361.2
XP_006716424.1
5
1881
XM_006716362.2
XP_006716425.1
4
1204
XM_006716363.1
XP_006716426.1
4
1376
XM_006716364.3
XP_006716427.1
3
1060
XM_011544578.1
XP_011542880.1
7
2253
XM_011544579.1
XP_011542881.1
6
2105
XM_011544580.1
XP_011542882.1
5
1983
XM_011544582.1
XP_011542884.1
5
1991
XM_011544583.1
XP_011542885.1
3
1170
XM_011544584.1
XP_011542886.1
3
1054
XM_011544585.2
XP_011542887.1
2
137
XM_017013623.1
XP_016869112.1
8
2303
XM_017013624.1
XP_016869113.1
5
1942
XM_017013625.1
XP_016869114.1
3
1219
XM_017013626.1
XP_016869115.1
4
1149
XM_017013627.1
XP_016869116.1
5
3543
XM_017013628.1
XP_016869117.1
4
3403
XM_017013629.1
XP_016869118.1
5
1334
XM_017013630.1
XP_016869119.1
5
1247
XM_017013631.1
XP_016869120.1
4
1208
XM_017013632.1
XP_016869121.1
5
1317
XM_017013633.1
XP_016869122.1
5
1304
XM_017013634.1
XP_016869123.1
5
1301
XM_017013635.1
XP_016869124.1
5
1309
XM_017013636.1
XP_016869125.1
6
1452
XM_017013637.1
XP_016869126.1
5
1309
XM_017013638.1
XP_016869127.1
6
1494
XM_017013639.1
XP_016869128.1
4
1142
GenBank mRNA
AB071403.1
-
2
696
AB081516.1
-
3
1014
AK055154.1
-
4
1831
AK314625.1
-
4
1085
AY358441.1
-
5
1326
BC060772.1
-
4
1131
30340553
BX649103.1
-
5
1686
Back To Top
More Information
Panther Classification:
Molecular Function -
glycosyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
endochondral ossification
nervous system development
cell recognition
cell proliferation
anatomical structure morphogenesis
heparan sulfate proteoglycan biosynthetic process, polysaccharide chain biosynthetic process
UDP-N-acetylgalactosamine metabolic process
proteoglycan biosynthetic process
extracellular matrix organization
chondroitin sulfate biosynthetic process
heparin biosynthetic process
UDP-glucuronate metabolic process
chondroitin sulfate proteoglycan biosynthetic process
dermatan sulfate proteoglycan biosynthetic process
chondroitin sulfate proteoglycan biosynthetic process, polysaccharide chain biosynthetic process
cartilage development
acetylgalactosaminyltransferase activity
peptidoglycan glycosyltransferase activity
glucuronosyltransferase activity
metal ion binding
glucuronylgalactosylproteoglycan 4-beta-N-acetylgalactosaminyltransferase activity
glucuronosyl-N-acetylgalactosaminyl-proteoglycan 4-beta-N-acetylgalactosaminyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline