Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CSGALNACT1 SIRNA Assays ›
          Gene Symbol:
          CSGALNACT1
          Gene Name:
          chondroitin sulfate N-acetylgalactosaminyltransferase 1
          Gene Aliases:
          CSGalNAcT-1, ChGn, beta4GalNAcT
          Chromosome Location:
          Chr.8: 19404161 - 19757940 on Build GRCh38
          UniGene ID:
          Hs.613729
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS125049
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CSGALNACT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130518.1 NP_001123990.1 9, 10 2381
          NM_018371.4 NP_060841.5 9, 10 1971
          NR_024040.1 9, 10 2050
          XM_006716358.2 XP_006716421.1 8, 9 2262
          XM_006716359.2 XP_006716422.1 9, 10 2631
          XM_006716360.2 XP_006716423.1 10, 11 2779
          XM_006716361.2 XP_006716424.1 9, 10 2526
          XM_006716362.2 XP_006716425.1 8, 9 1849
          XM_006716363.1 XP_006716426.1 8, 9 2021
          XM_006716364.3 XP_006716427.1 7, 8 1705
          XM_011544578.1 XP_011542880.1 11, 12 2898
          XM_011544579.1 XP_011542881.1 10, 11 2750
          XM_011544580.1 XP_011542882.1 9, 10 2628
          XM_011544582.1 XP_011542884.1 9, 10 2636
          XM_011544583.1 XP_011542885.1 7, 8 1815
          XM_011544584.1 XP_011542886.1 7, 8 1699
          XM_011544585.2 XP_011542887.1 6, 7 782
          XM_017013623.1 XP_016869112.1 12, 13 2948
          XM_017013624.1 XP_016869113.1 9, 10 2587
          XM_017013625.1 XP_016869114.1 7, 8 1864
          XM_017013626.1 XP_016869115.1 8, 9 1794
          XM_017013627.1 XP_016869116.1 9, 10 4188
          XM_017013628.1 XP_016869117.1 8, 9 4048
          XM_017013629.1 XP_016869118.1 9, 10 1979
          XM_017013630.1 XP_016869119.1 9, 10 1892
          XM_017013631.1 XP_016869120.1 8, 9 1853
          XM_017013632.1 XP_016869121.1 9, 10 1962
          XM_017013633.1 XP_016869122.1 9, 10 1949
          XM_017013634.1 XP_016869123.1 9, 10 1946
          XM_017013635.1 XP_016869124.1 9, 10 1954
          XM_017013636.1 XP_016869125.1 10, 11 2097
          XM_017013637.1 XP_016869126.1 9, 10 1954
          XM_017013638.1 XP_016869127.1 10, 11 2139
          XM_017013639.1 XP_016869128.1 8, 9 1787
          GenBank mRNA AB071403.1 - 6, 7 1341
          AB081516.1 - 7, 8 1659
          AK002126.1 - 6, 7 908
          AK314625.1 - 8, 9 1730
          AL157483.1 - 2, 3 178
          AY358441.1 - 9, 10 1971
          BC060772.1 - 9, 10 2063 30340553

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          endochondral ossification
          nervous system development
          cell recognition
          cell proliferation
          anatomical structure morphogenesis
          heparan sulfate proteoglycan biosynthetic process, polysaccharide chain biosynthetic process
          UDP-N-acetylgalactosamine metabolic process
          proteoglycan biosynthetic process
          extracellular matrix organization
          chondroitin sulfate biosynthetic process
          heparin biosynthetic process
          UDP-glucuronate metabolic process
          chondroitin sulfate proteoglycan biosynthetic process
          dermatan sulfate proteoglycan biosynthetic process
          chondroitin sulfate proteoglycan biosynthetic process, polysaccharide chain biosynthetic process
          cartilage development
          acetylgalactosaminyltransferase activity
          peptidoglycan glycosyltransferase activity
          glucuronosyltransferase activity
          metal ion binding
          glucuronylgalactosylproteoglycan 4-beta-N-acetylgalactosaminyltransferase activity
          glucuronosyl-N-acetylgalactosaminyl-proteoglycan 4-beta-N-acetylgalactosaminyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline