Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ALG13 SIRNA Assays ›
          Gene Symbol:
          ALG13
          Gene Name:
          ALG13, UDP-N-acetylglucosaminyltransferase subunit
          Gene Aliases:
          CDG1S, CXorf45, EIEE36, GLT28D1, MDS031, TDRD13, YGL047W
          Chromosome Location:
          Chr.X: 111680741 - 111764164 on Build GRCh38
          UniGene ID:
          Hs.443061
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS125164
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ALG13
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039210.4 NP_001034299.3 2 199
          NM_001099922.2 NP_001093392.1 2 199
          NM_001168385.2 NP_001161857.1 2 199
          NM_001257231.1 NP_001244160.1 2 272
          NM_001257237.1 NP_001244166.1 2 272
          NM_001257239.2 NP_001244168.1 2 187
          NM_001257240.2 NP_001244169.1 2 272
          NM_001257241.2 NP_001244170.1 2 272
          NM_001324290.1 NP_001311219.1 2 199
          NM_001324292.1 NP_001311221.1 2 199
          NM_001324293.1 NP_001311222.1 2 272
          NM_018466.5 NP_060936.1 2 199
          XM_006724693.2 XP_006724756.1 2 572
          XM_011531028.2 XP_011529330.1 2 573
          XM_011531030.1 XP_011529332.1 2 571
          XM_011531031.1 XP_011529333.1 2 272
          XM_011531033.2 XP_011529335.1 2 118
          XM_017029846.1 XP_016885335.1 2 574
          XM_017029847.1 XP_016885336.1 2 573
          XM_017029848.1 XP_016885337.1 2 571
          XM_017029849.1 XP_016885338.1 2 572
          XM_017029850.1 XP_016885339.1 2 571
          XM_017029851.1 XP_016885340.1 2 574
          GenBank mRNA AF220051.1 - 2 130
          AK302729.1 - 2 140
          AK302890.1 - 2 175
          AK304712.1 - 2 89
          AK311608.1 - 2 138
          AK312229.1 - 2 138
          AK316522.1 - 2 216
          BC005336.1 - 2 132 3996922
          BE747995.1 - 2 128
          BF239206.1 - 2 226
          BF699152.1 - 2 200
          BM759764.1 - 2 181
          DA830058.1 - 2 263
          DB292609.1 - 2 272
          DN992019.1 - 2 267

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cysteine protease transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          dolichol-linked oligosaccharide biosynthetic process
          proteolysis
          N-acetylglucosaminyldiphosphodolichol N-acetylglucosaminyltransferase activity
          thiol-dependent ubiquitinyl hydrolase activity
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-sskjb:80/100.66.75.127:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0