Search
Search

CMAS| Gene Symbol: CMAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018686.5 | NP_061156.1 | 5 | 845 | ||
| NR_135117.1 | 5 | 845 | ||||
| GenBank mRNA | AF271388.1 | - | 5 | 733 | ||
| AF397212.1 | - | 5 | 715 | |||
| AK222981.1 | - | 5 | 744 | |||
| AL832975.1 | - | 5 | 793 | |||
| BC016609.1 | - | 5 | 794 | 3448277 | ||