Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RFWD2 SIRNA Assays ›
          Gene Symbol:
          RFWD2
          Gene Name:
          ring finger and WD repeat domain 2
          Gene Aliases:
          COP1, RNF200
          Chromosome Location:
          Chr.1: 175944826 - 176207279 on Build GRCh38
          UniGene ID:
          Hs.523744
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS127525
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RFWD2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001740.3 NP_001001740.1 6 1040
          NM_001286644.1 NP_001273573.1 4 855
          NM_022457.6 NP_071902.2 6 1052
          XM_005245447.2 XP_005245504.1 6 1075
          XM_005245448.2 XP_005245505.1 6 1087
          XM_011509881.1 XP_011508183.1 6 1087
          XM_017002059.1 XP_016857548.1 6 1087
          XM_017002060.1 XP_016857549.1 6 1075
          XM_017002061.1 XP_016857550.1 6 1087
          XM_017002062.1 XP_016857551.1 6 1087
          XM_017002063.1 XP_016857552.1 6 1075
          XM_017002065.1 XP_016857554.1 6 1087
          XM_017002067.1 XP_016857556.1 6 1075
          XM_017002069.1 XP_016857558.1 3 792
          XM_017002070.1 XP_016857559.1 5 793
          XM_017002071.1 XP_016857560.1 3 182
          XM_017002072.1 XP_016857561.1 3 180
          XM_017002073.1 XP_016857562.1 3 182
          XM_017002074.1 XP_016857563.1 3 180
          XM_017002075.1 XP_016857564.1 4 316
          XM_017002076.1 XP_016857565.1 4 239
          XM_017002078.1 XP_016857567.1 6 441
          XM_017002079.1 XP_016857568.1 6 856
          XM_017002080.1 XP_016857569.1 2 95
          GenBank mRNA AF508940.1 - 6 786
          AF527539.1 - 6 1049
          AK001278.1 - 5 351
          AK314750.1 - 6 984
          AY509921.1 - 6 774
          BC039723.1 - 4 302
          BC094728.1 - 6 1040 5271629
          KJ534928.1 - 6 786
          KJ534929.1 - 6 786
          KJ534930.1 - 6 786
          KJ534932.1 - 6 774
          KJ534992.1 - 4 589
          KJ535061.1 - 3 491
          KJ535076.1 - 5 688

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to ionizing radiation
          protein ubiquitination
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          ligase activity
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0