Search
Search

SUSD1| Gene Symbol: SUSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282640.1 | NP_001269569.1 | 6 | 888 | ||
| NM_001282643.1 | NP_001269572.1 | 6 | 888 | |||
| NM_022486.4 | NP_071931.2 | 6 | 888 | |||
| XM_011518915.1 | XP_011517217.1 | 6 | 1111 | |||
| XM_011518916.2 | XP_011517218.1 | 6 | 1111 | |||
| XM_011518917.1 | XP_011517219.1 | 6 | 1111 | |||
| XM_011518918.1 | XP_011517220.1 | 6 | 1111 | |||
| XM_011518919.1 | XP_011517221.1 | 5 | 955 | |||
| XM_017015037.1 | XP_016870526.1 | 6 | 1111 | |||
| XM_017015038.1 | XP_016870527.1 | 6 | 1111 | |||
| XM_017015039.1 | XP_016870528.1 | 6 | 1111 | |||
| XM_017015040.1 | XP_016870529.1 | 6 | 1111 | |||
| GenBank mRNA | AK056704.1 | - | 6 | 776 | ||
| AK292868.1 | - | 6 | 888 | |||
| AL137432.1 | - | 2 | 295 | |||
| AY358746.1 | - | 6 | 867 | |||
| BC052314.1 | - | 2 | 139 | |||
| BC060770.1 | - | 6 | 858 | 30348765 | ||