Search
Search

CEP85| Gene Symbol: CEP85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281517.2 | NP_001268446.1 | 3 | 897 | ||
| NM_001319944.1 | NP_001306873.1 | 4 | 1050 | |||
| NM_022778.4 | NP_073615.2 | 4 | 1050 | |||
| XM_017002105.1 | XP_016857594.1 | 4 | 1050 | |||
| GenBank mRNA | AK024038.1 | - | 4 | 1006 | ||
| AK299312.1 | - | 3 | 850 | |||
| BC064528.1 | - | 4 | 1001 | 5585742 | ||