Search
Search

PINK1| Gene Symbol: PINK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032409.2 | NP_115785.1 | 4 | 1039 | ||
| GenBank mRNA | AB053323.1 | - | 4 | 1039 | ||
| AF316873.1 | - | 4 | 1015 | |||
| AK130746.1 | - | 2 | 1142 | |||
| BC009534.1 | - | 1 | 112 | |||
| BC028215.1 | - | 4 | 954 | 5214483 | ||