Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEMA6D SIRNA Assays ›
Gene Symbol:
SEMA6D
Gene Name:
semaphorin 6D
Gene Aliases:
-
Chromosome Location:
Chr.15: 47184101 - 47774223 on Build GRCh38
UniGene ID:
Hs.511265
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS129296
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEMA6D
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198999.1
NP_001185928.1
13
1268
NM_020858.1
NP_065909.1
10
1256
NM_024966.2
NP_079242.2
10
1256
NM_153616.1
NP_705869.1
10
1256
NM_153617.1
NP_705870.1
10
1256
NM_153618.1
NP_705871.1
10
1256
NM_153619.1
NP_705872.1
10
1256
XM_005254685.3
XP_005254742.1
10
1218
XM_005254686.2
XP_005254743.1
10
1162
XM_005254687.2
XP_005254744.1
10
1162
XM_005254689.3
XP_005254746.1
10
1218
XM_011522075.2
XP_011520377.1
13
1373
XM_011522076.2
XP_011520378.1
10
1162
XM_011522077.2
XP_011520379.1
10
1162
XM_011522078.2
XP_011520380.1
10
1162
XM_011522079.2
XP_011520381.1
10
1162
XM_011522080.2
XP_011520382.1
10
1162
XM_011522081.2
XP_011520383.1
10
1162
XM_017022617.1
XP_016878106.1
13
1186
XM_017022618.1
XP_016878107.1
10
1218
XM_017022619.1
XP_016878108.1
10
1218
XM_017022620.1
XP_016878109.1
13
1373
XM_017022621.1
XP_016878110.1
13
1373
GenBank mRNA
AB040912.2
-
13
1268
AF389426.1
-
10
1256
AF389427.1
-
10
1256
AF389428.1
-
10
1256
AF389429.1
-
10
1256
AF389430.1
-
10
1256
AK021660.1
-
7
554
AK290032.1
-
10
1177
BC150253.1
-
13
1268
9007179
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane-bound signaling molecule
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
neural crest cell migration
positive regulation of smooth muscle cell migration
negative regulation of smooth muscle cell migration
ventricular system development
positive regulation of cell migration
negative regulation of axon extension involved in axon guidance
negative chemotaxis
semaphorin-plexin signaling pathway
semaphorin receptor binding
chemorepellent activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0