Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NLRC5 SIRNA Assays ›
          Gene Symbol:
          NLRC5
          Gene Name:
          NLR family CARD domain containing 5
          Gene Aliases:
          CLR16.1, NOD27, NOD4
          Chromosome Location:
          Chr.16: 56989547 - 57083524 on Build GRCh38
          UniGene ID:
          Hs.528836
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS130675
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NLRC5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_032206.4 NP_115582.4 2 335
          XM_005256193.2 XP_005256250.1 3 396
          XM_005256194.3 XP_005256251.1 3 607
          XM_005256195.2 XP_005256252.1 3 396
          XM_005256196.2 XP_005256253.1 3 396
          XM_005256197.2 XP_005256254.1 3 396
          XM_005256199.3 XP_005256256.1 3 396
          XM_005256201.3 XP_005256258.1 3 396
          XM_006721297.2 XP_006721360.1 3 384
          XM_006721298.2 XP_006721361.1 3 396
          XM_006721300.2 XP_006721363.1 3 396
          XM_011523373.2 XP_011521675.1 3 543
          XM_011523375.2 XP_011521677.1 3 396
          XM_017023766.1 XP_016879255.1 3 396
          XM_017023767.1 XP_016879256.1 3 384
          XM_017023768.1 XP_016879257.1 3 396
          XM_017023769.1 XP_016879258.1 3 384
          XM_017023770.1 XP_016879259.1 3 396
          XM_017023771.1 XP_016879260.1 3 396
          GenBank mRNA AF389420.1 - 1 208
          AK074182.1 - 2 348
          AK097030.1 - 3 385
          AL704292.1 - 3 433
          DB293035.1 - 3 468
          EF452236.1 - 1 208

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-motor microtubule binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of NF-kappaB transcription factor activity
          negative regulation of type I interferon production
          regulation of kinase activity
          innate immune response
          positive regulation of MHC class I biosynthetic process
          positive regulation of transcription from RNA polymerase II promoter
          defense response to virus
          positive regulation of interferon-gamma-mediated signaling pathway
          negative regulation of type I interferon-mediated signaling pathway
          positive regulation of type I interferon-mediated signaling pathway
          RNA polymerase II core promoter sequence-specific DNA binding
          protein binding
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline