Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RHBDD1 SIRNA Assays ›
Gene Symbol:
RHBDD1
Gene Name:
rhomboid domain containing 1
Gene Aliases:
RRP4
Chromosome Location:
Chr.2: 226800164 - 226999210 on Build GRCh38
UniGene ID:
Hs.471514
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS130776
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RHBDD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001167608.1
NP_001161080.1
7, 8
1246
NM_032276.3
NP_115652.2
5, 6
945
XM_017005083.1
XP_016860572.1
7, 8
1240
XM_017005084.1
XP_016860573.1
7, 8
2315
XM_017005085.1
XP_016860574.1
6, 7
2301
XM_017005086.1
XP_016860575.1
6, 7
2233
XM_017005087.1
XP_016860576.1
5, 6
2082
XM_017005088.1
XP_016860577.1
6, 7
1021
XM_017005089.1
XP_016860578.1
7, 8
1260
XM_017005090.1
XP_016860579.1
6, 7
2096
XM_017005091.1
XP_016860580.1
6, 7
926
XM_017005092.1
XP_016860581.1
5, 6
2014
XM_017005093.1
XP_016860582.1
7, 8
1240
XM_017005094.1
XP_016860583.1
5, 6
953
XM_017005095.1
XP_016860584.1
7, 8
1240
GenBank mRNA
AK026955.1
-
3, 4
286
AK074258.1
-
5, 6
843
AL832364.1
-
4, 5
755
AX775825.1
-
5, 6
843
AY640233.1
-
4, 5
722
BC015553.1
-
3, 4
247
BC027900.1
-
2, 3
339
BC062636.1
-
2, 3
101
BC089404.1
-
2, 3
112
BC101262.1
-
4, 5
823
40023929
BC101263.1
-
4, 5
822
40023930
BC101264.1
-
4, 5
822
40023931
BC101265.1
-
4, 5
823
40023933
BC111056.1
-
5, 6
847
5748421
Back To Top
More Information
Panther Classification:
Molecular Function -
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
positive regulation of protein processing
membrane protein intracellular domain proteolysis
cellular response to unfolded protein
cellular response to UV
ERAD pathway
negative regulation of apoptotic process
post-translational protein modification
spermatid differentiation
membrane protein proteolysis involved in retrograde protein transport, ER to cytosol
regulation of male germ cell proliferation
serine-type endopeptidase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0