Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAPS2 SIRNA Assays ›
Gene Symbol:
CAPS2
Gene Name:
calcyphosine 2
Gene Aliases:
UG0636c06
Chromosome Location:
Chr.12: 75275979 - 75390941 on Build GRCh38
UniGene ID:
Hs.733835
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS131242
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAPS2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286547.1
NP_001273476.1
11
1157
NM_001286548.1
NP_001273477.1
13
1530
NM_001286549.1
NP_001273478.1
6
705
NM_032606.3
NP_115995.2
13
1402
XM_005269192.1
XP_005269249.1
11
1081
XM_005269194.3
XP_005269251.1
6
612
XM_006719648.2
XP_006719711.2
13
1402
XM_006719649.2
XP_006719712.2
12
1306
XM_011538878.1
XP_011537180.1
13
1367
XM_011538886.2
XP_011537188.1
12
1336
XM_011538887.2
XP_011537189.1
11
1240
XM_011538888.1
XP_011537190.1
9
921
XM_011538889.2
XP_011537191.1
13
1273
XM_011538890.1
XP_011537192.1
13
1402
XM_011538892.1
XP_011537194.1
10
967
XM_017020034.1
XP_016875523.1
13
1229
XM_017020035.1
XP_016875524.1
8
825
XM_017020036.1
XP_016875525.1
14
1492
XM_017020037.1
XP_016875526.1
11
1121
XM_017020038.1
XP_016875527.1
9
989
XM_017020039.1
XP_016875528.1
7
689
XM_017020040.1
XP_016875529.1
2
261
XM_017020041.1
XP_016875530.1
11
1227
XM_017020042.1
XP_016875531.1
11
1255
XM_017020043.1
XP_016875532.1
7
1072
XM_017020044.1
XP_016875533.1
6
666
XM_017020045.1
XP_016875534.1
7
1108
XM_017020046.1
XP_016875535.1
6
665
XM_017020047.1
XP_016875536.1
5
4118
GenBank mRNA
AF251056.1
-
13
1530
AF351611.1
-
12
1111
AK091839.1
-
13
1402
AK092274.1
-
7
618
BC033860.1
-
11
1126
BC056672.1
-
7
682
6616433
BC131724.1
-
6
704
Back To Top
More Information
Panther Classification:
Molecular Function -
calmodulin-related
Gene Ontology Categories:
Process(es)
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0