Search
Search

ATG4C| Gene Symbol: ATG4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032852.3 | NP_116241.2 | 4 | 436 | ||
| NM_178221.2 | NP_835739.1 | 4 | 388 | |||
| XM_005271288.2 | XP_005271345.1 | 4 | 379 | |||
| XM_011542313.2 | XP_011540615.1 | 4 | 410 | |||
| XM_017002600.1 | XP_016858089.1 | 4 | 410 | |||
| XM_017002601.1 | XP_016858090.1 | 4 | 388 | |||
| GenBank mRNA | AJ312234.1 | - | 3 | 241 | ||
| AJ320169.1 | - | 3 | 245 | |||
| AK027773.1 | - | 4 | 407 | |||
| BC008395.1 | - | 4 | 333 | |||
| BC018678.1 | - | 4 | 394 | |||
| BC033024.1 | - | 4 | 395 | 4838543 | ||
| BI518482.1 | - | 4 | 421 | |||
| DB183325.1 | - | 4 | 388 | |||