Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other N4BP2L2 SIRNA Assays ›
Gene Symbol:
N4BP2L2
Gene Name:
NEDD4 binding protein 2 like 2
Gene Aliases:
92M18.3, CG005, CG016, PFAAP5
Chromosome Location:
Chr.13: 32432488 - 32542710 on Build GRCh38
UniGene ID:
Hs.507680
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS131765
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
459.00
Your Price
Online offer:
459.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: N4BP2L2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001278432.1
NP_001265361.1
7
1698
NM_001320836.1
NP_001307765.1
7
3010
NM_033111.4
NP_149102.3
7
1653
XM_011534869.2
XP_011533171.1
6
2383
XM_011534870.2
XP_011533172.1
4
1389
XM_011534871.2
XP_011533173.1
4
1324
XM_011534872.2
XP_011533174.1
5
1511
XM_017020324.1
XP_016875813.1
8
2976
XM_017020325.1
XP_016875814.1
7
2970
XM_017020326.1
XP_016875815.1
7
2855
XM_017020327.1
XP_016875816.1
7
3003
XM_017020328.1
XP_016875817.1
8
2976
XM_017020329.1
XP_016875818.1
7
2970
XM_017020330.1
XP_016875819.1
7
3003
XM_017020331.1
XP_016875820.1
6
2258
XM_017020332.1
XP_016875821.1
7
2165
XM_017020333.1
XP_016875822.1
6
2258
XM_017020334.1
XP_016875823.1
7
2165
XM_017020335.1
XP_016875824.1
6
2143
XM_017020336.1
XP_016875825.1
4
1389
XM_017020337.1
XP_016875826.1
4
1324
XM_017020338.1
XP_016875827.1
4
1326
XM_017020339.1
XP_016875828.1
3
1646
XM_017020340.1
XP_016875829.1
4
1467
XM_017020341.1
XP_016875830.1
4
1440
XM_017020343.1
XP_016875832.1
3
1287
XM_017020344.1
XP_016875833.1
3
1203
XM_017020345.1
XP_016875834.1
4
1403
XM_017020346.1
XP_016875835.1
4
1394
GenBank mRNA
AK093514.1
-
4
1394
AK298546.1
-
6
1605
AK310280.1
-
4
1325
AL049802.1
-
5
1516
U50529.1
-
5
1517
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
positive regulation of hematopoietic stem cell proliferation
negative regulation of hematopoietic stem cell differentiation
RNA polymerase II transcription corepressor activity
molecular_function
protein binding
enzyme binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0