Search
Search

SIGLEC11| Gene Symbol: SIGLEC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135163.1 | NP_001128635.1 | 10 | 1861 | ||
| NM_052884.2 | NP_443116.2 | 11 | 2149 | |||
| XM_005258476.3 | XP_005258533.2 | 10 | 2042 | |||
| GenBank mRNA | AB072038.1 | - | 11 | 2068 | ||
| AF337818.1 | - | 11 | 2149 | |||
| AY358135.1 | - | 10 | 1739 | |||