Search
Search

ENPP6| Gene Symbol: ENPP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153343.3 | NP_699174.1 | 1, 2 | 394 | ||
| GenBank mRNA | AK057370.1 | - | 1, 2 | 339 | ||
| AY358676.1 | - | 1, 2 | 335 | |||
| BC035035.2 | - | 1, 2 | 299 | 5259487 | ||