Search
Search

TRIM50| Gene Symbol: TRIM50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281450.1 | NP_001268379.1 | 7 | 1476 | ||
| NM_001281451.1 | NP_001268380.1 | 7 | 1407 | |||
| NM_178125.3 | NP_835226.2 | 7 | 1479 | |||
| XM_011515788.2 | XP_011514090.1 | 6 | 1029 | |||
| XM_011515789.2 | XP_011514091.2 | 7 | 1404 | |||
| GenBank mRNA | AK057366.1 | - | 7 | 1477 | ||
| AK292074.1 | - | 7 | 1405 | |||
| AY081948.1 | - | 7 | 1479 | |||
| AY081949.1 | - | 7 | 1476 | |||
| BC112152.1 | - | 7 | 1321 | 8327620 | ||
| BC112154.1 | - | 7 | 1321 | 8327622 | ||
| BC143787.1 | - | 7 | 1318 | 9052301 | ||