Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other METTL21A SIRNA Assays ›
Gene Symbol:
METTL21A
Gene Name:
methyltransferase like 21A
Gene Aliases:
FAM119A, HCA557b, HSPA-KMT
Chromosome Location:
Chr.2: 207580631 - 207625928 on Build GRCh38
UniGene ID:
Hs.664764
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS135685
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: METTL21A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127395.2
NP_001120867.1
2
335
NM_001308021.1
NP_001294950.1
2
335
NM_145280.5
NP_660323.3
2
196
XM_005246336.3
XP_005246393.1
2
262
XM_005246337.4
XP_005246394.1
2
194
XM_005246338.4
XP_005246395.1
2
166
XM_005246339.3
XP_005246396.1
2
391
XM_005246340.3
XP_005246397.1
2
210
XM_005246341.2
XP_005246398.1
2
317
XM_006712327.3
XP_006712390.1
2
317
XM_006712329.3
XP_006712392.1
2
225
XM_011510724.2
XP_011509026.1
2
182
XM_011510725.2
XP_011509027.1
2
174
XM_011510727.2
XP_011509029.1
2
184
XM_011510728.2
XP_011509030.1
2
188
XM_011510729.2
XP_011509031.1
2
188
XM_017003442.1
XP_016858931.1
2
177
XM_017003443.1
XP_016858932.1
2
317
XM_017003444.1
XP_016858933.1
2
187
XM_017003445.1
XP_016858934.1
2
179
XM_017003446.1
XP_016858935.1
2
266
XM_017003447.1
XP_016858936.1
2
199
XM_017003448.1
XP_016858937.1
2
394
XM_017003449.1
XP_016858938.1
2
192
XM_017003450.1
XP_016858939.1
2
317
GenBank mRNA
AF455817.1
-
2
147
AK093812.1
-
2
196
BC009462.1
-
2
335
BC033720.1
-
2
207
4934087
BG832122.1
-
2
253
CN367990.1
-
2
134
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein methylation
peptidyl-lysine methylation
regulation of ATPase activity
protein binding
protein-lysine N-methyltransferase activity
Hsp70 protein binding
ATPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0