Search
Search

TMEM65| Gene Symbol: TMEM65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194291.2 | NP_919267.2 | 6 | 1129 | ||
| XM_011516847.2 | XP_011515149.1 | 6 | 1123 | |||
| XM_017013104.1 | XP_016868593.1 | 4 | 189 | |||
| GenBank mRNA | BC017881.1 | - | 4 | 233 | ||
| BC032396.1 | - | 6 | 430 | 5197382 | ||
| BC041379.1 | - | 6 | 1126 | 5275048 | ||