Search
Search

TMEM65| Gene Symbol: TMEM65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194291.2 | NP_919267.2 | 2, 3 | 882 | ||
| XM_011516847.2 | XP_011515149.1 | 2, 3 | 876 | |||
| GenBank mRNA | BC032396.1 | - | 2, 3 | 183 | 5197382 | |
| BC041379.1 | - | 2, 3 | 879 | 5275048 | ||