Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TPCN2 SIRNA Assays ›
          Gene Symbol:
          TPCN2
          Gene Name:
          two pore segment channel 2
          Gene Aliases:
          SHEP10, TPC2
          Chromosome Location:
          Chr.11: 69048876 - 69090604 on Build GRCh38
          UniGene ID:
          Hs.131851
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS137309
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TPCN2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_139075.3 NP_620714.2 22, 23 2123
          XM_005273824.3 XP_005273881.1 22, 23 2105
          XM_005273826.3 XP_005273883.1 20, 21 1816
          XM_005273830.3 XP_005273887.1 21, 22 1947
          XM_005273831.3 XP_005273888.1 19, 20 1799
          XM_005273832.3 XP_005273889.1 21, 22 2024
          XM_011544802.2 XP_011543104.1 21, 22 1996
          XM_011544807.2 XP_011543109.1 21, 22 1944
          XM_011544808.2 XP_011543110.1 19, 20 1874
          XM_017017328.1 XP_016872817.1 19, 20 1889
          XM_017017329.1 XP_016872818.1 19, 20 1886
          XM_017017330.1 XP_016872819.1 21, 22 2177
          XM_017017331.1 XP_016872820.1 19, 20 2347
          XM_017017332.1 XP_016872821.1 21, 22 2123
          XM_017017333.1 XP_016872822.1 18, 19 1807
          XM_017017334.1 XP_016872823.1 16, 17 1552
          XM_017017335.1 XP_016872824.1 15, 16 1487
          XM_017017336.1 XP_016872825.1 14, 15 1413
          GenBank mRNA AB209253.1 - 15, 16 1473
          AK023366.1 - 15, 16 1471
          AL137479.1 - 10, 11 866
          AW177543.1 - 3, 4 212
          AY029200.1 - 22, 23 2108
          BC063008.1 - 22, 23 2059 5214862

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ion channel transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular calcium ion homeostasis
          smooth muscle contraction
          lysosome organization
          regulation of autophagy
          calcium-mediated signaling
          response to vitamin D
          ion transmembrane transport
          regulation of ion transmembrane transport
          release of sequestered calcium ion into cytosol
          membrane depolarization during action potential
          voltage-gated calcium channel activity
          protein binding
          protein kinase binding
          identical protein binding
          NAADP-sensitive calcium-release channel activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0