Search
Search

HEPACAM2| Gene Symbol: HEPACAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039372.2 | NP_001034461.1 | 7, 8 | 1284 | ||
| NM_001288804.1 | NP_001275733.1 | 8, 9 | 1353 | |||
| NM_001288810.1 | NP_001275739.1 | 5, 6 | 1251 | |||
| NM_198151.2 | NP_937794.1 | 6, 7 | 1276 | |||
| XM_011516000.2 | XP_011514302.1 | 7, 8 | 1217 | |||
| XM_011516001.1 | XP_011514303.1 | 7, 8 | 1217 | |||
| GenBank mRNA | AK096002.1 | - | 7, 8 | 1237 | ||
| AK298361.1 | - | 8, 9 | 1303 | |||
| AY358345.1 | - | 6, 7 | 1238 | |||
| BC136300.1 | - | 6, 7 | 1206 | 9020287 | ||
| BC136301.1 | - | 6, 7 | 1206 | 9020288 | ||
| BC139906.1 | - | 6, 7 | 1263 | 40147445 | ||