Search
Search

MKX| Gene Symbol: MKX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242702.1 | NP_001229631.1 | 5 | 784 | ||
| NM_173576.2 | NP_775847.2 | 5 | 787 | |||
| XM_017016105.1 | XP_016871594.1 | 5 | 824 | |||
| XM_017016106.1 | XP_016871595.1 | 5 | 1015 | |||
| XM_017016107.1 | XP_016871596.1 | 4 | 692 | |||
| XM_017016108.1 | XP_016871597.1 | 4 | 692 | |||
| GenBank mRNA | AK125339.1 | - | 5 | 789 | ||
| BC036207.1 | - | 5 | 753 | 5262515 | ||