Search
Search

TMEM80| Gene Symbol: TMEM80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042463.2 | NP_001035928.2 | 4 | 412 | ||
| NM_001276274.1 | NP_001263203.1 | 4 | 412 | |||
| NM_174940.3 | NP_777600.3 | 4 | 388 | |||
| XM_006718206.3 | XP_006718269.3 | 4 | 418 | |||
| XM_017017600.1 | XP_016873089.1 | 4 | 593 | |||
| GenBank mRNA | AK124309.1 | - | 3 | 1454 | ||
| AK302585.1 | - | 4 | 207 | |||
| BC008671.1 | - | 4 | 336 | 3854028 | ||
| BI916171.1 | - | 4 | 390 | |||
| CF134831.1 | - | 3 | 178 | |||
| CK819789.1 | - | 4 | 202 | |||