Search
Search

IGSF10| Gene Symbol: IGSF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178822.4 | NP_849144.2 | 4 | 4284 | ||
| XM_011512708.2 | XP_011511010.1 | 10 | 5093 | |||
| XM_011512709.2 | XP_011511011.1 | 9 | 4824 | |||
| GenBank mRNA | AK128160.1 | - | 1 | 334 | ||
| AY273815.1 | - | 4 | 4284 | |||