Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NMNAT3 SIRNA Assays ›
Gene Symbol:
NMNAT3
Gene Name:
nicotinamide nucleotide adenylyltransferase 3
Gene Aliases:
FKSG76, PNAT-3, PNAT3
Chromosome Location:
Chr.3: 139560180 - 139678050 on Build GRCh38
UniGene ID:
Hs.208673
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS139491
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
459.00
Your Price
Online offer:
459.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NMNAT3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001200047.2
NP_001186976.1
4
888
NM_001320510.1
NP_001307439.1
7
1437
NM_001320511.1
NP_001307440.1
7
1189
NM_001320512.1
NP_001307441.1
6
1155
NM_001320513.1
NP_001307442.1
7
1567
NM_178177.4
NP_835471.1
5
1006
XM_011512790.1
XP_011511092.1
4
581
XM_011512791.2
XP_011511093.1
4
880
XM_017006328.1
XP_016861817.1
8
1454
XM_017006329.1
XP_016861818.1
8
1451
XM_017006330.1
XP_016861819.1
7
1414
XM_017006331.1
XP_016861820.1
8
1527
XM_017006332.1
XP_016861821.1
8
5468
XM_017006333.1
XP_016861822.1
6
1317
XM_017006334.1
XP_016861823.1
6
1314
XM_017006335.1
XP_016861824.1
7
1166
XM_017006336.1
XP_016861825.1
6
1126
XM_017006337.1
XP_016861826.1
5
1029
XM_017006338.1
XP_016861827.1
4
581
XM_017006339.1
XP_016861828.1
6
1269
XM_017006340.1
XP_016861829.1
5
1238
XM_017006341.1
XP_016861830.1
7
1197
XM_017006342.1
XP_016861831.1
8
1303
XM_017006343.1
XP_016861832.1
7
1263
XM_017006344.1
XP_016861833.1
6
1163
GenBank mRNA
AF345564.1
-
4
705
AK123208.1
-
4
803
AK127477.1
-
2
2991
BC034374.1
-
5
954
5185621
BC036218.1
-
7
1534
5270916
BX649063.1
-
2
1836
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotidyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
NAD biosynthetic process
response to wounding
NAD metabolic process
'de novo' NAD biosynthetic process from aspartate
nicotinamide-nucleotide adenylyltransferase activity
nicotinate-nucleotide adenylyltransferase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-blthd:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline