Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADM SIRNA Assays ›
          Gene Symbol:
          ADM
          Gene Name:
          adrenomedullin
          Gene Aliases:
          AM, PAMP
          Chromosome Location:
          Chr.11: 10304980 - 10307402 on Build GRCh38
          UniGene ID:
          Hs.441047
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS141163
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ADM
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001124.2 NP_001115.1 3 491
          GenBank mRNA AK312893.1 - 3 398
          BC015961.2 - 3 386 3506276
          BT006902.1 - 2 220
          CR541995.1 - 2 220
          D14874.1 - 3 376

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          peptide hormone intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          vasculogenesis
          response to hypoxia
          neural tube closure
          response to yeast
          regulation of the force of heart contraction
          G-protein coupled receptor internalization
          cAMP biosynthetic process
          progesterone biosynthetic process
          signal transduction
          positive regulation of cytosolic calcium ion concentration
          cell-cell signaling
          heart development
          female pregnancy
          aging
          blood circulation
          androgen metabolic process
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          response to cold
          response to wounding
          positive regulation of heart rate
          antibacterial humoral response
          antifungal humoral response
          cAMP-mediated signaling
          positive regulation of cAMP biosynthetic process
          organ regeneration
          neuron projection regeneration
          receptor internalization
          response to lipopolysaccharide
          response to insulin
          odontogenesis of dentin-containing tooth
          response to starvation
          positive regulation of apoptotic process
          negative regulation of vascular permeability
          positive regulation of angiogenesis
          negative regulation of vasoconstriction
          positive regulation of vasodilation
          hormone secretion
          developmental growth
          defense response to Gram-negative bacterium
          defense response to Gram-positive bacterium
          response to glucocorticoid
          branching involved in labyrinthine layer morphogenesis
          spongiotrophoblast layer development
          vascular smooth muscle cell development
          positive regulation of vasculogenesis
          receptor binding
          hormone activity
          protein binding
          adrenomedullin receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0