Search
Search

MNX1| Gene Symbol: MNX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165255.1 | NP_001158727.1 | 2 | 520 | ||
| NM_005515.3 | NP_005506.3 | 2 | 1076 | |||
| GenBank mRNA | AF107457.1 | - | 2 | 960 | ||
| AY927460.1 | - | 2 | 521 | |||
| AY927462.1 | - | 2 | 108 | |||