Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ID1 SIRNA Assays ›
          Gene Symbol:
          ID1
          Gene Name:
          inhibitor of DNA binding 1, HLH protein
          Gene Aliases:
          ID, bHLHb24
          Chromosome Location:
          Chr.20: 31605283 - 31606515 on Build GRCh38
          UniGene ID:
          Hs.504609
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS142591
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ID1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002165.3 NP_002156.2 1 279
          NM_181353.2 NP_851998.1 1 279
          GenBank mRNA AK291152.1 - 1 197
          AK313786.1 - 1 274
          BC000613.2 - 1 240 3346009
          BC012420.1 - 1 260 3862019
          BG546814.1 - 1 274
          BI768231.1 - 1 126
          BM856018.1 - 1 279
          BT007443.1 - 1 174
          D13889.1 - 1 192
          D13890.1 - 1 192
          DA839689.1 - 1 279
          S78825.1 - 1 184
          S78986.1 - 1 236
          X77956.1 - 1 206

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          angiogenesis
          endothelial cell morphogenesis
          transcription, DNA-templated
          transforming growth factor beta receptor signaling pathway
          brain development
          heart development
          circadian rhythm
          negative regulation of transcription by transcription factor localization
          positive regulation of gene expression
          neuron differentiation
          BMP signaling pathway
          protein destabilization
          positive regulation of actin filament bundle assembly
          collagen metabolic process
          cell-abiotic substrate adhesion
          negative regulation of apoptotic process
          regulation of MAPK cascade
          negative regulation of sequence-specific DNA binding transcription factor activity
          blood vessel endothelial cell migration
          negative regulation of endothelial cell differentiation
          negative regulation of osteoblast differentiation
          regulation of angiogenesis
          negative regulation of transcription, DNA-templated
          response to antibiotic
          blood vessel morphogenesis
          positive regulation of epithelial cell proliferation
          negative regulation of dendrite morphogenesis
          lung morphogenesis
          lung vasculature development
          cellular response to epidermal growth factor stimulus
          negative regulation of protein homodimerization activity
          cellular response to peptide
          negative regulation of RNA polymerase II regulatory region sequence-specific DNA binding
          cellular response to dopamine
          cellular response to nerve growth factor stimulus
          transcription factor activity, sequence-specific DNA binding
          protein binding
          protein C-terminus binding
          transcription factor binding
          protein homodimerization activity
          protein self-association
          protein N-terminus binding
          proteasome binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0