Search
Search

ID3| Gene Symbol: ID3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002167.4 | NP_002158.3 | 1 | 559 | ||
| GenBank mRNA | A17546.1 | - | 1 | 314 | ||
| A17548.1 | - | 1 | 191 | |||
| AK225965.1 | - | 1 | 294 | |||
| AK290003.1 | - | 1 | 296 | |||
| BC003107.1 | - | 1 | 277 | 3543936 | ||
| BM921819.1 | - | 590 | ||||
| BT006791.1 | - | 1 | 191 | |||
| X66924.1 | - | 1 | 314 | |||
| X69111.1 | - | 1 | 558 | |||