Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IRF5 SIRNA Assays ›
          Gene Symbol:
          IRF5
          Gene Name:
          interferon regulatory factor 5
          Gene Aliases:
          SLEB10
          Chromosome Location:
          Chr.7: 128937737 - 128950042 on Build GRCh38
          UniGene ID:
          Hs.521181
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS142677
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IRF5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098627.3 NP_001092097.2 3 564
          NM_001098629.2 NP_001092099.1 3 471
          NM_001098630.2 NP_001092100.1 3 471
          NM_001242452.2 NP_001229381.1 3 471
          NM_032643.4 NP_116032.1 3 387
          XM_005250317.3 XP_005250374.1 3 618
          XM_006715974.2 XP_006716037.1 3 554
          XM_011516158.2 XP_011514460.1 3 676
          XM_011516159.2 XP_011514461.1 3 585
          XM_011516160.1 XP_011514462.1 3 456
          XM_011516161.1 XP_011514463.1 3 486
          XM_011516162.1 XP_011514464.1 3 486
          XM_011516163.2 XP_011514465.1 3 672
          XM_011516164.1 XP_011514466.1 3 554
          GenBank mRNA AK297083.1 - 3 430
          AY504946.1 - 3 396
          AY504947.1 - 3 419
          AY693665.1 - 3 468
          AY693666.1 - 3 398
          AY693667.1 - 2 53
          AY693668.1 - 3 468
          AY693669.1 - 3 387
          BC004139.2 - 3 387 2963923
          BC004201.2 - 3 387 2963923
          BU540632.1 - 2 233
          BU950658.1 - 3 470
          DQ277633.1 - 3 387
          EU258895.1 - 2 224
          EU258896.1 - 3 379
          EU258897.1 - 3 478
          EU258898.1 - 3 562
          JQ950681.1 - 2 239
          JQ950682.1 - 1 89
          JQ950684.1 - 1, 2 163
          KP843527.1 - 3 447
          KP843528.1 - 3 447
          U51127.1 - 3 450

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001106586, XM_006236185, XM_006236180, XM_006236183, XM_006236184, and others.

          Panther Classification:

          Molecular Function -

          winged helix/forkhead transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          response to peptidoglycan
          response to muramyl dipeptide
          positive regulation of interferon-alpha production
          positive regulation of interferon-beta production
          positive regulation of interleukin-12 production
          positive regulation of apoptotic process
          positive regulation of transcription from RNA polymerase II promoter
          defense response to virus
          interferon-gamma-mediated signaling pathway
          type I interferon signaling pathway
          regulatory region DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0