Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MBD1 SIRNA Assays ›
          Gene Symbol:
          MBD1
          Gene Name:
          methyl-CpG binding domain protein 1
          Gene Aliases:
          CXXC3, PCM1, RFT
          Chromosome Location:
          Chr.18: 50266882 - 50281774 on Build GRCh38
          UniGene ID:
          Hs.405610
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS142881
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MBD1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001204136.1 NP_001191065.1 4 699
          NM_001204137.1 NP_001191066.1 4 699
          NM_001204138.1 NP_001191067.1 4 699
          NM_001204139.1 NP_001191068.1 4 699
          NM_001204140.1 NP_001191069.1 4 699
          NM_001204141.1 NP_001191070.1 4 699
          NM_001204142.1 NP_001191071.1 4 699
          NM_001204143.1 NP_001191072.1 4 480
          NM_001204151.2 NP_001191080.1 4 699
          NM_001323942.1 NP_001310871.1 4 699
          NM_001323947.1 NP_001310876.1 4 699
          NM_001323949.1 NP_001310878.1 4 691
          NM_001323950.1 NP_001310879.1 4 699
          NM_001323951.1 NP_001310880.1 4 699
          NM_001323952.1 NP_001310881.1 4 699
          NM_001323953.1 NP_001310882.1 4 706
          NM_001323954.1 NP_001310883.1 4 699
          NM_002384.2 NP_002375.1 4 699
          NM_015844.2 NP_056669.2 4 699
          NM_015845.3 NP_056670.2 4 699
          NM_015846.3 NP_056671.2 4 699
          NM_015847.3 NP_056723.2 4 699
          XM_005258271.2 XP_005258328.1 4 446
          XM_006722456.2 XP_006722519.1 4 446
          XM_011525991.1 XP_011524293.1 4 446
          XM_011525993.2 XP_011524295.1 4 446
          XM_011525994.2 XP_011524296.1 4 446
          XM_011525998.2 XP_011524300.1 4 446
          XM_011525999.1 XP_011524301.1 4 446
          XM_011526001.1 XP_011524303.1 4 446
          XM_011526002.1 XP_011524304.1 4 446
          XM_011526003.2 XP_011524305.1 4 446
          XM_011526006.1 XP_011524308.1 4 446
          XM_011526007.1 XP_011524309.1 4 446
          XM_017025751.1 XP_016881240.1 4 446
          XM_017025752.1 XP_016881241.1 4 446
          XM_017025753.1 XP_016881242.1 4 446
          XM_017025754.1 XP_016881243.1 4 446
          XM_017025755.1 XP_016881244.1 4 446
          XM_017025756.1 XP_016881245.1 4 446
          XM_017025757.1 XP_016881246.1 4 446
          XM_017025758.1 XP_016881247.1 4 446
          XM_017025759.1 XP_016881248.1 4 446
          XM_017025760.1 XP_016881249.1 4 446
          XM_017025761.1 XP_016881250.1 4 446
          XM_017025762.1 XP_016881251.1 4 446
          XM_017025763.1 XP_016881252.1 4 446
          XM_017025764.1 XP_016881253.1 4 446
          XM_017025765.1 XP_016881254.1 4 446
          XM_017025766.1 XP_016881255.1 4 446
          XM_017025767.1 XP_016881256.1 4 446
          XM_017025768.1 XP_016881257.1 4 446
          XM_017025769.1 XP_016881258.1 4 446
          XM_017025770.1 XP_016881259.1 4 446
          XM_017025771.1 XP_016881260.1 4 446
          XM_017025772.1 XP_016881261.1 4 446
          XM_017025773.1 XP_016881262.1 4 446
          XM_017025774.1 XP_016881263.1 4 446
          XM_017025775.1 XP_016881264.1 4 446
          XM_017025776.1 XP_016881265.1 4 446
          XM_017025777.1 XP_016881266.1 4 446
          GenBank mRNA AB209765.1 - 1 36
          AF078830.1 - 3 262
          AF078831.1 - 3 262
          AF078832.1 - 3 262
          AF078833.1 - 3 262
          AJ564845.1 - 4 401
          AK225889.1 - 4 609
          AK225948.1 - 4 405
          AK291519.1 - 4 700
          AK295401.1 - 4 431
          AK300759.1 - 4 480
          AK302004.1 - 4 391
          BC033242.1 - 4 425 5432943
          BC047353.1 - 4 384
          EF488685.1 - 3 262
          Y10746.1 - 4 401

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          methylation-dependent chromatin silencing
          transcription from RNA polymerase II promoter
          heart development
          aging
          response to fluoxetine
          neuron differentiation
          response to nutrient levels
          response to estradiol
          response to cocaine
          regulation of DNA methylation
          negative regulation of transcription, DNA-templated
          negative regulation of astrocyte differentiation
          DNA binding
          chromatin binding
          protein binding
          zinc ion binding
          methyl-CpG binding
          double-stranded methylated DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-jhh9h:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0