Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MBNL1 SIRNA Assays ›
Gene Symbol:
MBNL1
Gene Name:
muscleblind like splicing regulator 1
Gene Aliases:
EXP, MBNL
Chromosome Location:
Chr.3: 152243656 - 152465780 on Build GRCh38
UniGene ID:
Hs.201858
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS142882
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MBNL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001314057.1
NP_001300986.1
4
821
NM_021038.4
NP_066368.2
3
1606
NM_207292.2
NP_997175.1
3
1606
NM_207293.1
NP_997176.1
2
981
NM_207294.1
NP_997177.1
2
981
NM_207295.1
NP_997178.1
2
981
NM_207296.1
NP_997179.1
2
981
NM_207297.1
NP_997180.1
2
981
XM_005247457.4
XP_005247514.1
4
1174
XM_005247458.4
XP_005247515.1
4
2428
XM_005247460.4
XP_005247517.1
4
1137
XM_005247461.4
XP_005247518.1
4
1174
XM_005247462.4
XP_005247519.1
4
1174
XM_005247463.4
XP_005247520.1
4
1174
XM_005247464.4
XP_005247521.1
3
1232
XM_005247465.4
XP_005247522.1
4
1174
XM_005247466.4
XP_005247523.1
3
1209
XM_005247467.4
XP_005247524.1
4
1174
XM_005247468.4
XP_005247525.1
4
1174
XM_005247471.4
XP_005247528.1
3
1204
XM_005247474.3
XP_005247531.1
2
92
XM_005247475.3
XP_005247532.1
3
415
XM_005247476.4
XP_005247533.1
2
182
XM_005247477.4
XP_005247534.1
2
181
XM_006713639.1
XP_006713702.1
4
1156
XM_011512843.2
XP_011511145.1
4
1174
XM_011512845.2
XP_011511147.1
4
1174
XM_011512846.2
XP_011511148.1
4
1174
XM_011512847.2
XP_011511149.1
3
1137
XM_011512848.2
XP_011511150.1
4
778
XM_011512849.2
XP_011511151.1
3
1081
XM_017006423.1
XP_016861912.1
3
1425
XM_017006424.1
XP_016861913.1
3
1252
XM_017006425.1
XP_016861914.1
3
1254
XM_017006426.1
XP_016861915.1
3
1252
XM_017006427.1
XP_016861916.1
3
1399
XM_017006428.1
XP_016861917.1
3
1063
XM_017006429.1
XP_016861918.1
3
1146
XM_017006430.1
XP_016861919.1
3
1210
XM_017006431.1
XP_016861920.1
3
1287
XM_017006432.1
XP_016861921.1
3
1209
XM_017006433.1
XP_016861922.1
3
1209
XM_017006434.1
XP_016861923.1
3
1215
XM_017006435.1
XP_016861924.1
3
222
XM_017006436.1
XP_016861925.1
3
2335
XM_017006437.1
XP_016861926.1
3
1390
XM_017006438.1
XP_016861927.1
3
1081
XM_017006439.1
XP_016861928.1
3
415
XM_017006440.1
XP_016861929.1
2
149
XM_017006441.1
XP_016861930.1
2
182
XM_017006442.1
XP_016861931.1
2
285
XM_017006443.1
XP_016861932.1
2
293
GenBank mRNA
AB007888.1
-
3
1606
AF255334.1
-
2
703
AF395876.1
-
2
192
AF401998.1
-
2
192
AF497718.1
-
2
192
AF497719.1
-
2
192
AJ308400.1
-
2
221
BC043493.1
-
3
1111
6044671
BC050535.1
-
4
342
6042311
Y13829.1
-
2
703
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
mRNA processing
nervous system development
RNA splicing
embryonic limb morphogenesis
regulation of RNA splicing
myoblast differentiation
regulatory region RNA binding
RNA binding
double-stranded RNA binding
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0