Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PLAGL1 SIRNA Assays ›
          Gene Symbol:
          PLAGL1
          Gene Name:
          PLAG1 like zinc finger 1
          Gene Aliases:
          LOT1, ZAC, ZAC1
          Chromosome Location:
          Chr.6: 143940300 - 144064599 on Build GRCh38
          UniGene ID:
          Hs.743225
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS143317
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PLAGL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001080951.2 NP_001074420.1 8 951
          NM_001080952.2 NP_001074421.1 7 1103
          NM_001080953.2 NP_001074422.1 7 1057
          NM_001080954.2 NP_001074423.1 7 1056
          NM_001080955.2 NP_001074424.1 6 400
          NM_001080956.2 NP_001074425.1 6 595
          NM_001289037.1 NP_001275966.1 5 359
          NM_001289038.1 NP_001275967.1 4 313
          NM_001289039.1 NP_001275968.1 5 416
          NM_001289040.1 NP_001275969.1 6 385
          NM_001289041.1 NP_001275970.1 6 595
          NM_001289042.1 NP_001275971.1 5 747
          NM_001289043.1 NP_001275972.1 7 870
          NM_001289044.1 NP_001275973.1 6 821
          NM_001289045.1 NP_001275974.1 7 1365
          NM_001289046.1 NP_001275975.1 9 1346
          NM_001289047.1 NP_001275976.1 4 910
          NM_001289048.1 NP_001275977.1 7 1071
          NM_001289049.1 NP_001275978.1 6 1016
          NM_001317156.1 NP_001304085.1 4 1277
          NM_001317157.1 NP_001304086.1 3 1453
          NM_001317158.1 NP_001304087.1 3 815
          NM_001317159.1 NP_001304088.1 8 937
          NM_001317160.1 NP_001304089.1 6 581
          NM_001317161.1 NP_001304090.1 7 1071
          NM_001317162.1 NP_001304091.1 8 1097
          NM_006718.4 NP_006709.2 7 1089
          GenBank mRNA AJ006354.1 - 3 1483
          AJ303119.1 - 2 123
          AJ311395.1 - 9 1336
          AK091707.1 - 4 1277
          AK096331.1 - 7 1333
          AK314570.1 - 6 442
          AK315184.1 - 4 611
          BC074814.2 - 2 236 30915533
          BC109085.2 - 3 543 40034491
          BC109086.2 - 3 543 40034497
          BX537397.1 - 3 3784
          CR749329.1 - 2 4086
          DA426770.1 - 6 442
          DA549753.1 - 6 546
          DA675185.1 - 6 394
          DA717476.1 - 6 435
          U72621.4 - 6 2428
          U81992.1 - 3 207

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription from RNA polymerase II promoter
          apoptotic process
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          cell cycle arrest
          cell differentiation
          skeletal muscle cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II regulatory region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0