Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLAGL1 SIRNA Assays ›
Gene Symbol:
PLAGL1
Gene Name:
PLAG1 like zinc finger 1
Gene Aliases:
LOT1, ZAC, ZAC1
Chromosome Location:
Chr.6: 143940300 - 144064599 on Build GRCh38
UniGene ID:
Hs.743225
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS143318
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLAGL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080951.2
NP_001074420.1
8
983
NM_001080952.2
NP_001074421.1
7
1135
NM_001080953.2
NP_001074422.1
7
1089
NM_001080954.2
NP_001074423.1
7
1088
NM_001080955.2
NP_001074424.1
6
432
NM_001080956.2
NP_001074425.1
6
627
NM_001289037.1
NP_001275966.1
5
391
NM_001289038.1
NP_001275967.1
4
345
NM_001289039.1
NP_001275968.1
5
448
NM_001289040.1
NP_001275969.1
6
417
NM_001289041.1
NP_001275970.1
6
627
NM_001289042.1
NP_001275971.1
5
779
NM_001289043.1
NP_001275972.1
7
902
NM_001289044.1
NP_001275973.1
6
853
NM_001289045.1
NP_001275974.1
7
1397
NM_001289046.1
NP_001275975.1
9
1378
NM_001289047.1
NP_001275976.1
4
942
NM_001289048.1
NP_001275977.1
7
1103
NM_001289049.1
NP_001275978.1
6
1048
NM_001317156.1
NP_001304085.1
4
1309
NM_001317157.1
NP_001304086.1
3
1485
NM_001317158.1
NP_001304087.1
3
847
NM_001317159.1
NP_001304088.1
8
969
NM_001317160.1
NP_001304089.1
6
613
NM_001317161.1
NP_001304090.1
7
1103
NM_001317162.1
NP_001304091.1
8
1129
NM_006718.4
NP_006709.2
7
1121
GenBank mRNA
AJ006354.1
-
3
1515
AJ303119.1
-
2
155
AJ311395.1
-
9
1368
AK091707.1
-
4
1309
AK096331.1
-
7
1365
AK314570.1
-
6
474
AK315184.1
-
4
643
BC074814.2
-
2
268
30915533
BC109085.2
-
3
575
40034491
BC109086.2
-
3
575
40034497
BX537397.1
-
3
3816
CR749329.1
-
2
4118
DA426770.1
-
6
474
DA675185.1
-
6
426
DA717476.1
-
6
467
U72621.4
-
6
2460
U81992.1
-
3
239
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
apoptotic process
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
cell cycle arrest
cell differentiation
skeletal muscle cell differentiation
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0