Search
Search

SIX1| Gene Symbol: SIX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005982.3 | NP_005973.1 | 2 | 972 | ||
| XM_017021602.1 | XP_016877091.1 | 2 | 965 | |||
| GenBank mRNA | AK093780.1 | - | 2 | 1345 | ||
| BC008874.2 | - | 2 | 972 | 4138234 | ||
| BT007083.1 | - | 2 | 724 | |||
| X91868.1 | - | 2 | 999 | |||